Chick and Quail Brooder
Mother Hen Chick Brooder
Catalog # 470019-370
Supplier: HAWKEYE STEEL PRODUCTS INC
Fournisseur #: CQB20
Jump to:
Product Details & Documents
With a 10" interior height, this electric brooder can hold up to 125 day-old chicks, 50 month-old chicks, or 100 month-old quail chicks.
It has sliding mesh door panels on the front and one side, a 3 L poly bottle drinker and poly capacity feeder, and an easily removed dropping pan. It is constructed of galvanized steel and has a PVC coated wire floor. Some assembly is required. Thermostatic control is 250 W. 110 VAC.
Size: 27"W x 42"L x 19 1/2"H.
Weight: 67 lbs.
Spécifications
- Désignation:Mother Hen Chick Brooder