Chewing/Lapping Type, Honeybee Slide
Chewing/Lapping Type, Honeybee, Whole mount, Unstained
Catalog # 470181-640
Supplier:   Avantor
Chewing/Lapping Type, Honeybee Slide
Catalog # 470181-640
Supplier:  Avantor
Chewing/Lapping Type, Honeybee Slide
Chewing/Lapping Type, Honeybee Slide
Chewing/Lapping Type, Honeybee, Whole mount, Unstained
Price
$21.75
Log in pour la disponibilité
Each
Product Details & Documents
Demonstrates Insect Mandible Type
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Apis. Mouthparts are spread to show mandible, labial palp, maxilla, and labium. Unstained.
Spécifications
  • Désignation:
    Chewing/Lapping Type, Honeybee
  • Stain:
    Unstained
  • Orientation:
    Whole mount