Chewing/Lapping Type, Honeybee Slide
Chewing/Lapping Type, Honeybee, Whole mount, Unstained
Catalog # 470181-640
Supplier: Avantor
Jump to:
Product Details & Documents
Demonstrates Insect Mandible Type
- Identifying features clearly distinguishable
- Rigorous quality control standards
Apis. Mouthparts are spread to show mandible, labial palp, maxilla, and labium. Unstained.
Spécifications
- Désignation:Chewing/Lapping Type, Honeybee
- Stain:Unstained
- Orientation:Whole mount