Ward's® Chalcopyrite
Ward's® Chalcopyrite
Supplier:   Avantor
Ward's® Chalcopyrite
Supplier:  Avantor
Certificates
Ward's® Chalcopyrite
Ward's® Chalcopyrite
3 Options Available Below
Product Details & Documents
Good grade, brass-yellow copper ore, massive.
  • Collected in Mexico
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25 to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Informations sur le produit
  • Désignation
    Specimen Dimensions
    Availability
    Price
  • Chalcopyrite, student specimens
    1×1 - 1×1"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $22.39
    $22.39
    Pack of 10
       $22.39
  • Chalcopyrite, hand specimen
    2×2 - 3×4"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $67.15
    $67.15
    Each
       $67.15
  • Chalcopyrite, bulk pack, 1 kg
    Size will vary
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $60.19
    $60.19
    Each
       $60.19
Storage
Informations sur le produit
  • Catalog #
    Désignation
    Availability
    Price
  • Storage box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2.35
    $2.35
    Each
       $2.35