Certified pH Buffer Set | Avantor
Certified pH Buffer Set
Kit, Certified pH Buffer Set, GLOBE Approved Product
Catalog # 34175-485
Supplier:   Micro Essential
Fournisseur #:  CC-471V
Certified pH Buffer Set
Catalog # 34175-485
Supplier:  Micro Essential
CAS Number:  
Certified pH Buffer Set
Certified pH Buffer Set
Kit, Certified pH Buffer Set, GLOBE Approved Product
CAS Number:  
Price
$67.36
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Set includes calibrated and pre-weighed amounts of buffer powders at three pH values (pH 4.0, 7.0, and 10.0) for testing or calibrating your pH meter.
Powders come in capsules and each is sufficient for 100 ml of buffer solution.
Delivery information: Includes 10 capsules for each pH value and a vial of buffer preservative.
Spécifications
Spécifications
  • Désignation:
    Kit, Certified pH Buffer Set, GLOBE Approved Product