ContactUsLinkComponent
Preserved Cat or Dog Flea
Catalog # 470001-798
Supplier:  Avantor
undefined
Preserved Cat or Dog Flea
Catalog # 470001-798
Supplier:  Avantor
Specifications
  • Description:
    Cat or Dog Flea, Alcohol Preserved
  • Size:
    1/8"
  • Injection Type:
    Uninjected
  • Cat. no.:
    470001-798
  • Packaging:
    Vail of 10
  • Supplier No.:
    WARD470001-798
Specifications
About this item
Alcohol preserved fleas. Available in vials.
  • Entomology
  • Invertebrate anatomy
Ctenocephalides sp. Specialized hind legs allow the flea to jump as high as 13". Size: 1/8".