Capillaria hepatica Slide
Capillaria hepatica, Section, Hematoxylin and Eosin
Catalog # 470182-176
Supplier: Avantor
Jump to:
Product Details & Documents
Parasitic Nematode
- Identifying features clearly distinguishable
- Rigorous quality control standards
Liver worm. Trichurid nematode parasite of a variety of animals and humans. Liver showing ova.
Spécifications
- Désignation:Capillaria hepatica
- Stain:Hematoxylin and Eosin
- Orientation:Section