Foam Calorimeter Set | Avantor
Foam Calorimeter Set
Calorimeter Set, Foam
Catalog # 470012-650
Supplier:   Avantor
Foam Calorimeter Set
Catalog # 470012-650
Supplier:  Avantor
Foam Calorimeter Set
Foam Calorimeter Set
Calorimeter Set, Foam
Price
$100.30
Log in pour la disponibilité
Each Pallet of 60
Product Details & Documents
Ensure your studies of heat exchange are safe, easy, and fun.
These economical calorimeters features slotted lids to allow insertion of a thermometer to take measutremnts without losing heat to the surrounding air. Each holds 500 ml of water.
Delivery information: Includes a set of 10 calorimeters and instructions.
Spécifications
  • Désignation:
    Calorimeter Set, Foam