ContactUsLinkComponent
Bugula, Colony Slide
Catalogue # 470181-564
Fournisseur:  Avantor
undefined
Bugula, Colony Slide
Catalogue # 470181-564
Fournisseur:  Avantor
Spécifications
  • Description:
    Bugula, Colony
  • Cat. no.:
    470181-564
  • Stain:
    Schneider's
  • Supplier No.:
    WARD470181-564
  • Orientation:
    Whole mount
Spécifications
À propos de cet article
Marine Bryozoan
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Portion of colony. Commonly mistaken for seaweed.