Electrophoresis Running Buffer
Electrophoresis Running Buffer
Buffer
Catalog # 470111-864
Supplier:   Edvotek
Supplier #:  607
Electrophoresis Running Buffer
Catalog # 470111-864
Supplier:  Edvotek
CAS Number:  
Electrophoresis Running Buffer
Electrophoresis Running Buffer
Buffer
CAS Number:  
Price
$65.75
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Tris Acetate, pH 7.8, concentrated 50X (for 5 L—20 standard gels).
Specifications
Specifications
  • Désignation:
    Buffer
Specifications