Bridge Rectifier | Avantor
Bridge Rectifier
Bridge Rectifier
Catalog # 470121-246
Supplier:   Wards
Fournisseur #:  PH 1134-N8
Bridge Rectifier
Catalog # 470121-246
Supplier:  Wards
Bridge Rectifier
Bridge Rectifier
Bridge Rectifier
Price
$99.80
Log in pour la disponibilité
Each
Product Details & Documents
This Bridge Rectifier is a must for current studies.
  • Rectifier Converts AC Input to DC Output
  • Four Diodes Connect Inside a Bridge Circuit
  • Clear Circuit Diagram
  • Students Learn to Rectify Alternating Current to Direct Current
  • Purpose for Students: to Discover the Importance of Diodes
Four diodes connected in a bridge circuit can be used to convert alternating current (AC) input into direct current (DC) output.
The unit features a clear circuit diagram as well, so students see the importance of diodes and how to rectify alternating current to direct current.
Spécifications
  • Désignation:
    Bridge Rectifier