Bottle Rocket Launcher | Avantor
Bottle Rocket Launcher
Supplier:   Science First
Bottle Rocket Launcher
Supplier:  Science First
Certificates
Bottle Rocket Launcher
2 Options Available Below
Product Details & Documents
A Safe and Easy Alternative to Engine Rockets
Take your students outside to show them a dramatic demonstration of the laws of motion. With the simple materials in the bottle rocket kit, any standard plastic soda bottle that you provide up to two liters in size, can be launched into the air. It comes with operating instructions and suggested experiments. Stand size: 8"L x 2"W x 1"H, Length of air line: 5'; length of trigger line: 10'.
Note: Base launcher not included.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Dual Bottle Rocket Launcher
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 187,40 $
    187,40 $
    Each
       187,40 $
    Supplier Number: 17010
  • Single Bottle Rocket Launcher
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 96,15 $
    96,15 $
    Each
       96,15 $
    Supplier Number: 17000
Accessories
Informations sur le produit
  • Catalog #
    Height
    Availability
    Price
  • Bicycle Pump with Ø 11/4"
    Bicycle Pump with Ø 11/4"
    470089-326
    17¼"
    Log in pour la disponibilité
    Each - $80.80