Bone, Cancellous Slide
Bone, Cancellous, Cross section, Hematoxylin and Eosin
Catalog # 470177-584
Supplier: Avantor
Jump to:
Product Details & Documents
Explore spongy bone that produces red blood cells
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Decalcified and stained.
Spécifications
- Désignation:Bone, Cancellous
- Stain:Hematoxylin and Eosin
- Orientation:Cross section