Bone Development Slide
Bone development slide, Cross section, Mallory Triple
Catalog # 470182-658
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates Components of Maturing Bone
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Femur of newborn or young mammal.
Spécifications
- Désignation:Bone development slide
- Stain:Mallory Triple
- Orientation:Cross section