Bone Cutting Shears
Bone Cutting Shears
Supplier:   Dunrite Instruments
Bone Cutting Shears
Certificates
Bone Cutting Shears
Bone Cutting Shears
3 Options Available Below
Product Details & Documents
Makes easy work of cutting bone.
  • Flush cutting face
  • Spring handles
  • Stainless steel construction wont chip
  • Stainless construction
Make one of the toughest dissection chores — cutting through bone — easier with straight shears designed especially for the job. These shears will also spare wear and tear on your fine instruments.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Product Information
  • Désignation
    Longueur
    Matière
    Availability
    Price
  • Bone Cutting Shears
    130 mm (5.1")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $56.95
    $56.95
    Each
       $56.95
    Supplier Number: 140995
  • Bone Cutting Shears
    190 mm (7.5")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $83.05
    $83.05
    Each
       $83.05
    Supplier Number: 140997
  • Bone Cutting Shears
    222 mm (8.75")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $92.25
    $92.25
    Each
       $92.25
    Supplier Number: 141110