Blunt-Point Surgical Scissors
Blunt-Point Surgical Scissors, 140 mm (5.5"), Stainless steel, Blunt/Blunt
Catalog # 470018-946
Supplier: Dunrite Instruments
Fournisseur #: 140985
Jump to:
Product Details & Documents
When working with particularly delicate specimens, the blunt tips of these straight scissors reduce the chance of tearing, damaging, or puncturing tissue adjacent to the cut. Stainless steel.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
- Désignation:Blunt-Point Surgical Scissors
- Longueur:140 mm (5.5")
- Matière:Stainless steel
- Blade Type:Blunt/Blunt