Blocks and Refraction Plates
Blocks and Refraction Plates
Blocks and Refraction Plates
Blocks and Refraction Plates
Parallel-Sided Blocks for Ray Optics
Ideal for refraction and internal reflection studies as well as index of refraction measurements, these clear blocks are free of bubbles and streaks and have polished surfaces. Acrylic blocks and refraction plates have frosted ends, glass block is completely clear.
Product Family Options
  • Désignation
  • Blocks and Refraction Plates, Clear Glass Block, 114 x 63 x 19mm
    Each
    Each - $18.95
    $18.95
    Each
       $18.95
     : PH0566D
  • Blocks and Refraction Plates, Acrylic Block, 79 x 27 x13mm
    Each
    Each - $18.95
    $18.95
    Each
       $18.95
     : PH0568-13
  • Blocks and Refraction Plates, Acrylic Block, 100 x 75 x 20mm
    Each
    Each - $18.95
    $18.95
    Each
       $18.95
     : PH0568E
  • Blocks and Refraction Plates, Acrylic Block, 70 x 70 x 9mm
    Each
    Each - $19.85
    $19.85
    Each
       $19.85
     : PH0568E
  • Blocks and Refraction Plates, Glass Refraction Plate, 70 x 70 x 6mm
    Each
    Each - $17.89
    $17.89
    Each
       $17.89
     : 1-58