Ward's® Biotite
Biotite Thin Section
  470024-938
 :   Avantor
Ward's® Biotite
  470024-938
 :  Avantor
Ward's® Biotite
Ward's® Biotite
Biotite Thin Section
$96.59
Each
When minerals, rocks, and fossils cannot be accurately identified by macroscopic observation and testing, thin section slides can reveal the composition and structure of the specimens on a microscopic level.
Thin section slides are precisely ground to a thickness of 0.03 mm across the entire specimen area. Mineral and rock thin sections are mounted on 27 x 46 mm microscope slides, and protected by a glass coverslip. All thin section slides are labeled for reference.
Our excellent series of standard thin sections features important and interesting rock-forming mineral types most often found in field samples.
  • Désignation:
    Biotite Thin Section