ContactUsLinkComponent
Biological Diversity - Three Domains Slide Set
Catalogue # 470183-414
Fournisseur:  Avantor
undefined
Biological Diversity - Three Domains Slide Set
Catalogue # 470183-414
Fournisseur:  Avantor
Spécifications
  • Type:
    Cellular Development
  • Description:
    Biological Diversity - Three Domains Slide Set
  • No. of Slides:
    3
  • Cat. no.:
    470183-414
  • Supplier No.:
    WARD470183-414
Spécifications
À propos de cet article
Signifies Evolutionary Split of Cellular Structure
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Investigate three domains of simple organisms.
Ordering information: Set includes Archaea: Archaebacteria, mixed types (sm); Prokarya (Bacteria): Bacteria forms, mixed smears (sm); and Eukarya: Mixed protists (wm).