Biological Diversity, Eight Kingdoms Slide Set
470183-418
: Avantor
- Type:Cellular Development
- Description:Biological Diversity, Eight Kingdoms Slide Set
- No. of Slides:8
- Cat. no.:470183-418
- Supplier No.:WARD470183-418
Alternate System That Seeks to Resolve Some Traditional Taxonomic Problems
- Identifying features clearly distinguishable
- Rigorous quality control standards
All are prepared as wholemounts unless otherwise noted.
: Sets include Archaea: Archaebacteria, mixed types (sm); Bacteria: Bacillus (sm); Fungi: Saccharomyces; Archaezoa: Giardia lamblia; Protista: Paramecium, four types; Chromista: Ectocarpus; Plantae: Fern, Sporophyte; Animalia: Amphioxus.