ContactUsLinkComponent
Biological Diversity, Eight Kingdoms Slide Set
  470183-418
 :  Avantor
undefined
Biological Diversity, Eight Kingdoms Slide Set
  470183-418
 :  Avantor
  • Type:
    Cellular Development
  • Description:
    Biological Diversity, Eight Kingdoms Slide Set
  • No. of Slides:
    8
  • Cat. no.:
    470183-418
  • Supplier No.:
    WARD470183-418
Alternate System That Seeks to Resolve Some Traditional Taxonomic Problems
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
All are prepared as wholemounts unless otherwise noted.
 : Sets include Archaea: Archaebacteria, mixed types (sm); Bacteria: Bacillus (sm); Fungi: Saccharomyces; Archaezoa: Giardia lamblia; Protista: Paramecium, four types; Chromista: Ectocarpus; Plantae: Fern, Sporophyte; Animalia: Amphioxus.