ContactUsLinkComponent
Biofuels: Investigating Ethanol Production and Combustion Lab Activity
Catalog # 470024-534
Supplier:  Lab-Aids
undefined
Biofuels: Investigating Ethanol Production and Combustion Lab Activity
Catalog # 470024-534
Supplier:  Lab-Aids
Supplier Number:  39S
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Biofuels: Investigating Ethanol Production and Combustion Lab Activity
  • No. of Lab Groups:
    6
  • Cat. no.:
    470024-534
  • Supplier No.:
    39S-A
Specifications
About this item
Demonstrate the Costs and Benefits of Biofuels
  • Grades 6-12
  • Produce energy from readily available materials
  • Appproximate classtime: 3 periods of 50 min each
In three phases of investigation students will gain understanding of alternative fuels, how they are made, what they are used for, and what trade-offs exist. Students begin by fermenting corn and cane sugar, the two food sources used most commonly to produce ethanol. The experiment continues with the distillation of the fermented stillage to produce fuel and studying the energy released by the combustion of ethanol and kerosene. Comparing these results and constrasting the two by-products — CO2 and particulate matter — of the two fuels, students can then discuss the trade-offs of biofuels and fossil fuels as sources of energy. Includes materials for six setups.