Digital Timer
Digital timer, 99 min, 59 s, 6.5×5.5×1.5 cm
Catalog # 470093-772
Supplier: Taylor Precision Products
Fournisseur #: 5806
Jump to:
Product Details & Documents
The multi-purpose digital timer is versatile and easy to use.
- Small and compact design
- Counts up and counts down
- Clip/magnet/stand positioning
This lightweight timer can count up (stopwatch function) or count down. A 0.7 LCD readout provides a clear display that can be read from a distance. The timer is capable of programming up to 99 minutes.
Power supply: AAA battery
Packaging: Retail card.
Spécifications
- Désignation:Digital timer
- W×H×D:6.5×5.5×1.5 cm
- Minuterie:99 min, 59 s