Benchtop DNA Marker
Catalog # 470180-822
Supplier: PROMEGA SUNNYVALE INC SE
CAS Number:
Specifications
- Description:Benchtop DNA Marker, 300 μL
- Cat. no.:470180-822
- Supplier No.:G8291
Specifications
About this item
Ready to use, this DNA marker does not require preparation or refrigeration — it can be used effectively at room temperature. For additional convenience, the marker is supplied in blue/orange loading dye, so you can tract migration during electrophoresis. Load 6 μL per well. Total quantity: 300 μL.