ContactUsLinkComponent
Benchtop DNA Marker
Catalog # 470180-822
CAS Number:  
undefined
Benchtop DNA Marker
Catalog # 470180-822
Supplier Number:  G8291
CAS Number:  
Specifications
  • Description:
    Benchtop DNA Marker, 300 μL
  • Cat. no.:
    470180-822
  • Supplier No.:
    G8291
Specifications
About this item
Ready to use, this DNA marker does not require preparation or refrigeration — it can be used effectively at room temperature. For additional convenience, the marker is supplied in blue/orange loading dye, so you can tract migration during electrophoresis. Load 6 μL per well. Total quantity: 300 μL.