Basic Soil Sampler Set
Basic Soil Sampler Set, 8", 1"
  470004-344
 :   Avantor
Basic Soil Sampler Set
  470004-344
 :  Avantor
Basic Soil Sampler Set
Basic Soil Sampler Set
Basic Soil Sampler Set, 8", 1"
$144.99
KIT(1items) Kit of 1 Pallet of 48
Get Perfect Samples in Any Kind of Soil
  • Remove Your Core Intact
  • Easy to Use in the Field
  • Teacher Guide Included
Use the boring handle to select your sample, then just remove it and replace it with a special extractor unit. This special handle neatly and easily removes your soil plug intact. The sampler also features a depth measurement scale, allowing you to collect multiple samples from uniform depths up to 8" with a minimum of fuss. When you are finished in the field, just use the boring handle or a stiff brush to clean it. It comes with enough soil collection bags and labels for 30 students, a teacher’s guide, and a student study guide copymaster.
  • Désignation:
    Basic Soil Sampler Set
  • Longueur:
    8"
  • Diameter:
    1"