Basic Plant Slide Set
Basic Plant Slide Set, 12, Plant
Catalog # 470183-422
Supplier: Avantor
Jump to:
Product Details & Documents
Explore the Form and Structure of Various Plant Tissues
- Identifying features clearly distinguishable
- Rigorous quality control standards
Ordering information: Set includes Mixed pollen, 16 types (wm); Monocot root tip (wm); Allium (onion), epidermis (wm); Lillium (lily), young flower bud (cs); Zea, (corn), root tip (ls); Zea, stem (cs); Zea, leaf (cs); Zea, kernel and embryo (ls); Capsella (shepherd’s purse), mature embryos (ls); Ligustrum (privet), leaf (cs); Ranunculus (buttercup), mature root (cs); and Tilia (basswood), 3-year stem (cs).
Specifications
- Type:Plant
- Désignation:Basic Plant Slide Set
- No. of Slides:12