Ward's® Basic Microbiological Techniques Lab Activity
Ward's® Basic Microbiological Techniques Lab Activity
Basic Microbiological Techniques Kit, 6
Catalog # 470030-196
Supplier:   Avantor
Ward's® Basic Microbiological Techniques Lab Activity
Catalog # 470030-196
Supplier:  Avantor
Ward's® Basic Microbiological Techniques Lab Activity
Ward's® Basic Microbiological Techniques Lab Activity
Basic Microbiological Techniques Kit, 6
Price
$521.40
Log in pour la disponibilité
Kit of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Learn How to Identify Bacteria Using Several Different Staining Procedures
  • Demonstrates multiple techniques
  • Includes two bacterial cultures
  • Materials for six setups
  • Time Required: ~ 90 minutes
By employing culturing and staining techniques including sterile transfer, streak plating, Gram staining, and endospore staining, students identify the included “unknown” bacteria. These procedures also familiarize students with sterile technique and preparation of microscope slides. Tube cultures: Bacillus cereus and Escherichia coli. Includes a teacher’s guide, and student copymaster.
Canada: Containment Level 2 – PHAC License required.
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.
Spécifications
  • Désignation:
    Basic Microbiological Techniques Kit
  • No. of Lab Groups:
    6