Ward's® Bacti-Spreader
Bacti-Spreader, 9", 2"
Catalog # 470004-860
Supplier: Dunrite Instruments
Fournisseur #: 151993
Jump to:
Product Details & Documents
Special Design Evenly Distributes Bacteria
- Reusable for multiple inoculations
The stainless steel rod has been specifically designed for greater contact with agar for more even distribution of bacteria.
Spécifications
- Désignation:Bacti-Spreader
- Longueur:9"
- Blade Width:2"