Bacteria Survey Slide Set
Catalogue # 470183-438
Fournisseur: Avantor
Spécifications
- Type:Monera
- Description:Bacteria Survey Slide Set
- No. of Slides:7
- Cat. no.:470183-438
- Supplier No.:WARD470183-438
Spécifications
À propos de cet article
Phylogenetic Examples of Different Morphological Types and Metabolic Strategies
- All identifying features are clearly distinguishable
- Rigorous quality control standards
- In-house experts available to answer your questions
Explore the many morphological differences and specific characteristics of different bacteria types. All preparations are smears unless otherwise noted.
Ordering information: Set includes Archae; Archaebacteria, mixed types; Spirochete: Spirochaete sp.; Gram Positive: Clostridium; Cyanobacteria; Anabaena (wm); Proteobacteria-Chemoheterotroph: Salmonella; Chemoautotroph: Rhizobium; and Phototroph: Chromobacterium.