Bacillus anthracis, Gram (+) Slide
Bacillus anthracis Slide, Smear, Gram (+)
470180-888
: Avantor
Learn about bacterial morphology and infections.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Rods. It causes anthrax in humans and animals.
- Désignation:Bacillus anthracis Slide
- Stain:Gram (+)
- Orientation:Smear