Atomic Mobiles Activity
470152-836
: Avantor
- Description:Atomic Mobiles Activity
- No. of Lab Groups:19
- Cat. no.:470152-836
- Includes:Instructions
- Supplier No.:WARD470152-836
Students gain a better understanding of the structure of matter.
- Chemistry Art for the Classroom
- Hands-On Depiction of Elements
- Students Gain a Better Understanding of the Structure of Matter
- Periodic Table Included
- Detailed Instruction Manual Included
Not only do they love to build these mobiles, but they’ll also be able to hang their work from the ceiling for display.
The hands-on structure of this activity make it easy to comprehend the elements.
: This set includes 74 wooden spheres in nine colors representing nuclei and electrons; 19 large rings, scaled to represent the covalent radii of elements from hydrogen to potassium; a periodic table; an assortment of fine monofilament and small parts to assemble and suspend the models; and a detailed instruction manual.