<i>Arthrobotrys conoides</i>
Arthrobotrys conoides
Arthrobotrys conoides, Cornmeal agar, 25 °C, Tube
Catalog # 470179-634
Supplier:   Avantor
<i>Arthrobotrys conoides</i>
Catalog # 470179-634
Supplier:  Avantor
<i>Arthrobotrys conoides</i>
Arthrobotrys conoides
Arthrobotrys conoides, Cornmeal agar, 25 °C, Tube
Price
$20.45
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Deuteromycete. Biological control organism. Traps nematodes. Incubation temperature 25°C. Cornmeal agar growth medium.
Specifications
  • Désignation:
    Arthrobotrys conoides
  • Packaging:
    Tube
  • Media:
    Cornmeal agar
  • Incubation Conditions:
    25 °C