Arsenic Quick Test Kit | Avantor
Arsenic Quick Test Kit
Arsenic Quick Test Kit
Catalog # 470021-304
Fournisseur #:  481396
Arsenic Quick Test Kit
Catalog # 470021-304
Arsenic Quick Test Kit
Arsenic Quick Test Kit
Arsenic Quick Test Kit
Price
$434.55
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Safe, Accurate, and Cost-Effective Test for Arsenic in Water or Soil
This colorimetric arsenic test only takes 12 minutes to complete, and it will detect a range of 0–500 ppb. The kit comes with 100 tests.
Spécifications
  • Désignation:
    Arsenic Quick Test Kit