Ancient Microbes, Mixed Types Slide
Ancient microbes, mixed types, Smear, Gram (+/-)
Catalog # 470176-986
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates old type bacteria/Archaea.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Rods/cocci, involuted forms. Mixture of various types of 'ancient bacteria' representing extreme halophiles (Halobacterium salinarium (rods), Staphylococcus saprophyticus (coccus), and Cellumonas sp. (rods)).
Spécifications
- Désignation:Ancient microbes, mixed types
- Stain:Gram (+/-)
- Orientation:Smear