ContactUsLinkComponent
Aqua Scope II Underwater Viewer
Aqua Scope II Underwater Viewer
Catalog # 470006-624
Supplier:  Science First
undefined
Aqua Scope II Underwater Viewer
Catalog # 470006-624
Supplier:  Science First
Supplier Number:  623-8110
Specifications
  • Description:
    AquaVue Underwater Viewer
  • Length:
    26"
  • Cat. no.:
    470006-624
  • Supplier No.:
    78-015
Specifications
About this item
For consistent readings underwater.
  • Clear viewing window
    Lightweight, durable
    Simple and convenient operation
The AquaVue Underwater Viewer offers a clear, immersive view of aquatic life and environments, even under challenging conditions. Its innovative design eliminates surface glare and internal reflections, ensuring consistent visibility regardless of lighting or weather. Ideal for educational use, it allows students to explore underwater ecosystems during field trips and outdoor biology lessons. The lightweight, crush-resistant viewer includes a flotation collar, neoprene viewing mask, and handle for easy handling, and is also effective for precise Secchi disk readings. Built for durability and repeated use, it comes with instructions for quick setup and operation.