Antibiotic Sensitivity and Bacteria Activity Kit
Catalog # 470149-980
Supplier: G-Biosciences
Specifications
- Description:Antibiotic Sensitivity & Bacteria Activity Kit
- Cat. no.:470149-980
- Supplier No.:BE-203
Specifications
About this item
Teach the Importance of Aseptic Techniques
Demonstrate the concepts of antibiotic selection, bacteria sensitivity and resistance, and applications for screening for antibiotic resistance with this hands-on kit. Students will determine which antibiotic is the most suitable to treat a bacterial infection by analyzing the inhibitory effects of antibiotics on selected bacteria cells. Materials supplied for six groups of up to five students.