Two-Chamber Animal Behavior Tray
Catalogue # 470019-336
Fournisseur: Avantor
Spécifications
- Description:Animal Behavior Trays, 2-Chamber
- Cat. no.:470019-336
- Supplier No.:WARD470019-336
Spécifications
À propos de cet article
Design your own experiments in animal behavior, or use them to expand an existing kit.
- Easily cleaned for reuse
- Trays will nest for easy compact storage
Each of these formed plastic trays can be used for simple taxis experiments in small organisms.
It is a set of 10