Analog Stopwatch | Avantor
Analog Stopwatch
Analog stopwatch
Catalog # 470123-440
Supplier:   Wards
Fournisseur #:  PH0358A
Analog Stopwatch
Catalog # 470123-440
Supplier:  Wards
Analog Stopwatch
Analog Stopwatch
Analog stopwatch
Liquidation
$97.00
Vente
Valable jusqu'au 31/12/2026
Log in pour la disponibilité
Each
Product Details & Documents
Count up to 29:59 with resolution of ¹/₅ sec. It is nickel-plated and features two-dials and a hard carrying case.
Size: 2"Ø.
Spécifications
  • Désignation:
    Analog stopwatch