Anabaena Slide
Catalog # 470176-974
Supplier: Avantor
Specifications
- Description:Anabaena
- Cat. no.:470176-974
- Stain:Safranin
- Supplier No.:WARD470176-974
- Orientation:Whole Mount
Specifications
About this item
Illustrates the characteristic chain of cells.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Filaments with heterocysts and spores, showing chains of cells.