Amphioxus, Composite Slide
Amphioxus, Composite, Cross section, Mallory Triple
Catalog # 470181-690
Supplier: Avantor
Jump to:
Product Details & Documents
Lancelet Study
- Identifying features clearly distinguishable
- Rigorous quality control standards
Cross section through the buccal funnel, anterior pharynx, gonads, intestine, and tail.
Spécifications
- Désignation:Amphioxus, Composite
- Stain:Mallory Triple
- Orientation:Cross section