Amphibole Set
Amphibole Set
470024-864
: Avantor
Specially selected thin sections of important rock forming minerals. Set may contain more than one example of the same mineral to depict variations in occurrence and distinct mineral associations.
Slides (12): actinolite, anthophyllite, cummingtonite, ferrohornblende, fluoredenite, glaucophane, grunerite, hexagonite, kaersutite, magnesiohornblende, riebeckite, tremolite.
- Désignation:Amphibole Set