Amoeba Proteus Routine Slide, Explano-Mount™
Catalogue # 470183-642
Fournisseur: Avantor
Spécifications
- Description:Amoeba proteus, Routine Explano-Mount™
- Cat. no.:470183-642
- Stain:Trichrome
- Supplier No.:WARD470183-642
- Orientation:Explano-Mount™
Spécifications
À propos de cet article
Slide and Illustrated Guide Packed in a Durable Plastic Sleeve
- Identifying features clearly distinguishable
- Rigorous quality control standards
Routine preparation, numerous specimens.