Ammonia Test Kit | Avantor
Ammonia Test Kit
Ammonia Aquarium Test Kit
Catalog # 470021-372
Supplier:   Pet World Inc
Fournisseur #:  LR8600
Ammonia Test Kit
Catalog # 470021-372
Supplier:  Pet World Inc
Ammonia Test Kit
Ammonia Test Kit
Ammonia Aquarium Test Kit
Price
$34.00
Log in pour la disponibilité
Kit of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Fast and easy-to-use, this kit contains two dropper bottles for accurate dispensing of test solutions, non-staining test tube with snap cap, and instructions. It comes with enough solution for 130 tests and can be used in either fresh or salt water. Range: 0–8 ppm ammonia.
Spécifications
  • Désignation:
    Ammonia Aquarium Test Kit