Order Entry
Canada
ContactUsLinkComponent
Amino Acid Set
Catalog # 470300-170
Supplier:  Ward's Science
CAS Number:  
undefined
Amino Acid Set
Catalog # 470300-170
Supplier:  Ward's Science
Supplier Number:  AA0042-KIT
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Amino Acids Set
  • Size:
    Set of 22
  • Chemical/Physical Specifications:
    1 g Each
  • Cat. no.:
    470300-170
  • Grade:
    Laboratory
  • Supplier No.:
    AA0042-KIT

Specifications

About this item

Shelf Life (months): 12
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.