Alligator Cords
Alligator cord, 56 cm (22")
Catalog # 470112-724
Supplier: Elenco Electronics
Fournisseur #: TL6-LG
Jump to:
Product Details & Documents
Standard Test Leads.
- Set of 10 cords
Colors include red, black, yellow, white, and green.
Spécifications
- Désignation:Alligator cord
- Cord Length:56 cm (22")