Algae Types Slides
Product Details & Documents
Varying examples of algae classifications
- Identifying features clearly distinguishable
- Rigorous quality control standards
This selection of slides is designed to supplement lessons found in most high school and introductory college textbooks. The representative selection allows students to study the classification of algae - a controversial topic due to the fact that algae may be grouped in as many as three separate kingdoms. All slides are whole mounts except for Fucus monoecious, which is a section.
Ordering information: Set includes Chlamydomonas(wm); Cladophora(wm); Desmids(wm); Oedogonium(wm); Pediastrum(wm); Spirogyra conjugation(wm); Blue-green algae(wm); Batrachospermum(wm); Volvox(wm); Euglena(wm); Diatoms(wm); Fucus monoecious (sec).
Spécifications
- Type:Plant
- Désignation:Algae Types Slides
- No. of Slides:12