Ward's® Algae Types Collection | Avantor
Ward's® Algae Types Collection
WARD’S Algae Types Collection
Catalog # 470000-954
Supplier:   Avantor
Ward's® Algae Types Collection
Catalog # 470000-954
Supplier:  Avantor
Ward's® Algae Types Collection
Ward's® Algae Types Collection
WARD’S Algae Types Collection
Price
53,65 $
Log in pour la disponibilité
Kit of 1
Product Details & Documents
Formalin preserved.
Includes blue-green algae from the kingdom Monera, along with green, brown, and red algae from the kingdom Protista. Supplied in separate, labeled jars. Set of four.
Spécifications
  • Désignation:
    WARD’S Algae Types Collection
Accessories
Informations sur le produit
  • Catalog #
    Grade
    Size
    Availability
    Price
  • WARDSafe™
    WARDSafe™
    470233-546
    Laboratory
    19 L
    Log in pour la disponibilité
    Each (19l) - $345.85