ContactUsLinkComponent
Alconox® Detergent Powder
Alconox® Detergent Powder
Catalog # 470200-838
Supplier:  Alconox
undefined
Alconox® Detergent Powder
Catalog # 470200-838
Supplier:  Alconox
Supplier Number:  1104
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Alconox® Detergent Powder, 1.8 kg (4lb)
  • Cat. no.:
    470200-838
  • Supplier No.:
    1104
Specifications
About this item
Scientifically Formulated for Manual Cleaning.
  • Mild (pH 9-9.5) anionic powder detergent
  • Biodegradable
Performs equally well in hard or soft, cold or hot water. It leaves no residue after rinsing. Everything cleaned becomes bright, shiny and sparkling.