Ward's® Albite
Ward's® Albite
Ward's® Albite
Certificates
Ward's® Albite
5 Options Available Below
Product Details & Documents
Feldspar group; white to gray, sodium plagioclase cleavages; some faces may show twinning.
  • Collected in Ontario, Canada
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25 to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Informations sur le produit
  • Désignation
    Specimen Dimensions
    Availability
    Price
  • Albite, hand specimen, each
    2×3 - 3×4"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $33.50
    $33.50
    Each
       $33.50
    Spécifications:
    More
    Specifications
    Cat. No.
    470015-414
  • Albite thin section
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $96.59
    $96.59
    Each
       $96.59
    Spécifications:
    More
    Specifications
    Cat. No.
    470024-880
  • Albite, bulk pack, 1 kg
    Size will vary
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $34.90
    $34.90
    Each
       $34.90
    Spécifications:
    More
    Specifications
    Cat. No.
    470025-394
  • Albite, student specimens
    1×1 - 1×1"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $33.80
    $33.80
    Pack of 10
       $33.80
    Spécifications:
    More
    Specifications
    Cat. No.
    470015-412
Storage
Informations sur le produit
  • Désignation
    Specimen Dimensions
    Availability
    Price
  • Storage box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2.35
    $2.35
    Each
       $2.35
    Supplier Number: 196883
    Spécifications:
    More
    Specifications
    Cat. No.
    470147-278
Storage
Informations sur le produit
  • Catalog #
    Désignation
    Availability
    Price
  • Storage box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2.35
    $2.35
    Each
       $2.35