Prepared Agarose
Supplier:   Ward's Science
Prepared Agarose
Supplier:  Ward's Science
Certificates
Prepared Agarose
Prepared Agarose
2 Options Available Below
Product Details & Documents
Prepared with TBE buffer, these gels are easy to use — simply melt them in a microwave oven or water bath then pour them into a casting tray. The non-reusable gels have a one-year shelf life and should be stored at room temperature.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Prepared Agarose Gel, 0.8%, 200 ml Bottle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (200ml)
    Each (200ml) - $33.00
    $33.00
    Each (200ml)
       $33.00
    Supplier Number: AA0084-200ML
  • Prepared Agarose Gel, 2.0%, 200 ml Bottle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (200ml)
    Each (200ml) - $40.90
    $40.90
    Each (200ml)
       $40.90
    Supplier Number: AA0082-200ML