Advanced Plant Slide Set
Advanced Plant Slide Set, 100, Plant
Catalog # 470183-486
Supplier: Avantor
Jump to:
Product Details & Documents
Study Cellular and Tissue Organization of Plant Anatomy and Phylogenetic Comparison
- Identifying features clearly distinguishable
- Rigorous quality control standards
Ligustrum and Nymphaea leaves showing the variation in structure.
Ordering information: Set Includes 100 slides allowing an in-depth exploration of the diverse plant kingdom. Slides include the following types of slides from different species: protective wood fibers, stems; vascular system; root system; reproductive structures; pollen and embryos.
Specifications
- Type:Plant
- Désignation:Advanced Plant Slide Set
- No. of Slides:100