Advanced Leaf Classification Riker Mount
Advanced Leaf Classification Riker Mount, 20 x 14"
Catalog # 470200-894
Supplier: WHITE OWL ENTERPRISES
Supplier #: R102
Jump to:
Product Details & Documents
Cut down on the time spent teaching students how to identify leaves by margin and structure variations.
This display of carefully pressed, labeled specimens makes it easy.
It measures 14 x 20".
Spécifications
- Désignation:Advanced Leaf Classification Riker Mount
- Taille:20 x 14"